Anti-CA2 Rat Monoclonal Antibody [clone: 322706]

Supplier: R & D Systems
MAB2184
220019-661EA 594.37 USD
220019-661
Anti-CA2 Rat Monoclonal Antibody [clone: 322706]
Antibodies
Type: Primary
Antigen: CA2
Clonality: Monoclonal
Clone: 322706
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR