Anti-CA1 Rat Monoclonal Antibody [clone: 363121]

Supplier: R & D Systems
MAB2180
220019-660EA 577.02 USD
220019-660
Anti-CA1 Rat Monoclonal Antibody [clone: 363121]
Antibodies
Type: Primary
Antigen: CA1
Clonality: Monoclonal
Clone: 363121
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR