Anti-IL11 Mouse Monoclonal Antibody [clone: 22626]

Supplier: R & D Systems
MAB218
220019-659EA 779.48 USD
220019-659
Anti-IL11 Mouse Monoclonal Antibody [clone: 22626]
Antibodies
Type: Primary
Antigen: IL11
Clonality: Monoclonal
Clone: 22626
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR