Anti-TNFSF18 Rat Monoclonal Antibody [clone: 721926]

Supplier: R & D Systems
MAB21772
102885-440EA 594.37 USD
102885-440
Anti-TNFSF18 Rat Monoclonal Antibody [clone: 721926]
Antibodies
Type: Primary
Antigen: TNFSF18
Clonality: Monoclonal
Clone: 721926
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR