Anti-CTSB Mouse Monoclonal Antibody [clone: 155714]

Supplier: R & D Systems
MAB2176
220019-655EA 772.25 USD
220019-655
Anti-CTSB Mouse Monoclonal Antibody [clone: 155714]
Antibodies
Type: Primary
Antigen: CTSB
Clonality: Monoclonal
Clone: 155714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR