Anti-SERPINE2 Rat Monoclonal Antibody [clone: 244404]

Supplier: R & D Systems
MAB2175
220019-654EA 763.57 USD
220019-654
Anti-SERPINE2 Rat Monoclonal Antibody [clone: 244404]
Antibodies
Type: Primary
Antigen: SERPINE2
Clonality: Monoclonal
Clone: 244404
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR