Anti-STAT5A Mouse Monoclonal Antibody [clone: 251619]

Supplier: R & D Systems
MAB2174
220019-652EA 600.15 USD
220019-652
Anti-STAT5A Mouse Monoclonal Antibody [clone: 251619]
Antibodies
Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 251619
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR