Anti-STAT6 Rat Monoclonal Antibody [clone: 69727]

Supplier: R & D Systems
MAB2169
220019-648EA 647.88 USD
220019-648
Anti-STAT6 Rat Monoclonal Antibody [clone: 69727]
Antibodies
Type: Primary
Antigen: STAT6
Clonality: Monoclonal
Clone: 69727
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR