Anti-PTX3 Rat Monoclonal Antibody [clone: 265641]

Supplier: R & D Systems
MAB21661
220019-646EA 772.25 USD
220019-646
Anti-PTX3 Rat Monoclonal Antibody [clone: 265641]
Antibodies
Type: Primary
Antigen: PTX3
Clonality: Monoclonal
Clone: 265641
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR