Anti-PTX3 Rat Monoclonal Antibody [clone: 265629]

Supplier: R & D Systems
MAB2166
220019-645EA 594.37 USD
220019-645
Anti-PTX3 Rat Monoclonal Antibody [clone: 265629]
Antibodies
Type: Primary
Antigen: PTX3
Clonality: Monoclonal
Clone: 265629
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR