Anti-CXCR4 Rat Monoclonal Antibody [clone: 247506]

Supplier: R & D Systems
MAB21651-100
102885-438EA 655.11 USD
102885-438
Anti-CXCR4 Rat Monoclonal Antibody [clone: 247506]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 247506
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR