Anti-CXCR2 Rat Monoclonal Antibody [clone: 242216]

Supplier: R & D Systems
MAB2164-100
102885-436EA 655.11 USD
102885-436
Anti-CXCR2 Rat Monoclonal Antibody [clone: 242216]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 242216
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR