Anti-CSF1 Mouse Monoclonal Antibody [clone: 26786]

Supplier: R & D Systems
MAB2161
220019-642EA 763.57 USD
220019-642
Anti-CSF1 Mouse Monoclonal Antibody [clone: 26786]
Antibodies
Type: Primary
Antigen: CSF1
Clonality: Monoclonal
Clone: 26786
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR