Anti-VTCN1 Rat Monoclonal Antibody [clone: 297219]

Supplier: R & D Systems
MAB2154
220019-638EA 633.42 USD
220019-638
Anti-VTCN1 Rat Monoclonal Antibody [clone: 297219]
Antibodies
Type: Primary
Antigen: VTCN1
Clonality: Monoclonal
Clone: 297219
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR