Anti-HC Rat Monoclonal Antibody [clone: 295103]

Supplier: R & D Systems
MAB2150
220019-636EA 577.02 USD
220019-636
Anti-HC Rat Monoclonal Antibody [clone: 295103]
Antibodies
Type: Primary
Antigen Symbol: HC
Antigen Name: hemolytic complement
Antigen Synonyms: C5, C5a, He
Clonality: Monoclonal
Clone: 295103
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR