Anti-CHL1 Rat Monoclonal Antibody [clone: 316316]

Supplier: R & D Systems
MAB2147
220019-633EA 600.15 USD
220019-633
Anti-CHL1 Rat Monoclonal Antibody [clone: 316316]
Antibodies
Type: Primary
Antigen: CHL1
Clonality: Monoclonal
Clone: 316316
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR