Anti-CAMP Mouse Monoclonal Antibody [clone: 250532]

Supplier: R & D Systems
MAB2146
220019-632EA 772.25 USD
220019-632
Anti-CAMP Mouse Monoclonal Antibody [clone: 250532]
Antibodies
Type: Primary
Antigen: CAMP
Clonality: Monoclonal
Clone: 250532
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR