Anti-CXCR6 Rat Monoclonal Antibody [clone: 221002]

Supplier: R & D Systems
MAB2145-100
102885-432EA 633.42 USD
102885-432
Anti-CXCR6 Rat Monoclonal Antibody [clone: 221002]
Antibodies
Type: Primary
Antigen: CXCR6
Clonality: Monoclonal
Clone: 221002
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR