Anti-LAMA3 Mouse Monoclonal Antibody [clone: 546215]

Supplier: R & D Systems
MAB21441
102885-430EA 647.88 USD
102885-430
Anti-LAMA3 Mouse Monoclonal Antibody [clone: 546215]
Antibodies
Type: Primary
Antigen: LAMA3
Clonality: Monoclonal
Clone: 546215
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR