Anti-LAMA3 Mouse Monoclonal Antibody [clone: P3H9-2]

Supplier: R & D Systems
MAB2144
220019-630EA 584.25 USD
220019-630
Anti-LAMA3 Mouse Monoclonal Antibody [clone: P3H9-2]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: LAMA3
Clonality: Monoclonal
Clone: P3H9-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR