Anti-VASN Mouse Monoclonal Antibody [clone: 227526]

Supplier: R & D Systems
MAB2140
220019-629EA 779.48 USD
220019-629
Anti-VASN Mouse Monoclonal Antibody [clone: 227526]
Antibodies
Type: Primary
Antigen: VASN
Clonality: Monoclonal
Clone: 227526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR