Anti-LAMC1 Mouse Monoclonal Antibody [clone: D18]

Supplier: R & D Systems
MAB2139
220019-627EA 600.15 USD
220019-627
Anti-LAMC1 Mouse Monoclonal Antibody [clone: D18]
Antibodies
Type: Primary
Antigen: LAMC1
Clonality: Monoclonal
Clone: D18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR