Anti-CCL2 Mouse Monoclonal Antibody [clone: 238607]

Supplier: R & D Systems
MAB2137
220019-625EA 763.57 USD
220019-625
Anti-CCL2 Mouse Monoclonal Antibody [clone: 238607]
Antibodies
Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 238607
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR