Anti-WFIKKN1 Mouse Monoclonal Antibody [clone: 289819]

Supplier: R & D Systems
MAB2136
220019-622EA 561.11 USD
220019-622
Anti-WFIKKN1 Mouse Monoclonal Antibody [clone: 289819]
Antibodies
Type: Primary
Antigen: WFIKKN1
Clonality: Monoclonal
Clone: 289819
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR