Anti-TNFRSF6B Mouse Monoclonal Antibody [clone: 229812]

Supplier: R & D Systems
MAB2135
220019-620EA 772.25 USD
220019-620
Anti-TNFRSF6B Mouse Monoclonal Antibody [clone: 229812]
Antibodies
Type: Primary
Antigen: TNFRSF6B
Clonality: Monoclonal
Clone: 229812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR