Anti-IL9R Rat Monoclonal Antibody [clone: 224325]

Supplier: R & D Systems
MAB2134
220019-619EA 763.57 USD
220019-619
Anti-IL9R Rat Monoclonal Antibody [clone: 224325]
Antibodies
Type: Primary
Antigen: IL9R
Clonality: Monoclonal
Clone: 224325
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR