Anti-SIGLEC10 Mouse Monoclonal Antibody [clone: 265817]

Supplier: R & D Systems
MAB2130
220019-616EA 600.15 USD
220019-616
Anti-SIGLEC10 Mouse Monoclonal Antibody [clone: 265817]
Antibodies
Type: Primary
Antigen: SIGLEC10
Clonality: Monoclonal
Clone: 265817
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR