Anti-ADAMTS17 Mouse Monoclonal Antibody [clone: 136133]

Supplier: R & D Systems
MAB21291
220019-614EA 763.57 USD
220019-614
Anti-ADAMTS17 Mouse Monoclonal Antibody [clone: 136133]
Antibodies
Type: Primary
Antigen: ADAMTS17
Clonality: Monoclonal
Clone: 136133
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR