Anti-ADAMTS17 Mouse Monoclonal Antibody [clone: 136121]

Supplier: R & D Systems
MAB2129
220019-613EA 772.25 USD
220019-613
Anti-ADAMTS17 Mouse Monoclonal Antibody [clone: 136121]
Antibodies
Type: Primary
Antigen: ADAMTS17
Clonality: Monoclonal
Clone: 136121
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR