Anti-KREMEN1 Mouse Monoclonal Antibody [clone: 301324]

Supplier: R & D Systems
MAB2127
220019-612EA 577.02 USD
220019-612
Anti-KREMEN1 Mouse Monoclonal Antibody [clone: 301324]
Antibodies
Type: Primary
Antigen: KREMEN1
Clonality: Monoclonal
Clone: 301324
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR