Anti-CHL1 Rat Monoclonal Antibody [clone: 316223]

Supplier: R & D Systems
MAB2126
220019-611EA 600.15 USD
220019-611
Anti-CHL1 Rat Monoclonal Antibody [clone: 316223]
Antibodies
Type: Primary
Antigen: CHL1
Clonality: Monoclonal
Clone: 316223
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR