Anti-IGFBP7 Rat Monoclonal Antibody [clone: 292917]

Supplier: R & D Systems
MAB21201
220019-609EA 577.02 USD
220019-609
Anti-IGFBP7 Rat Monoclonal Antibody [clone: 292917]
Antibodies
Type: Primary
Antigen: IGFBP7
Clonality: Monoclonal
Clone: 292917
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR