Anti-GPC3 Mouse Monoclonal Antibody [clone: 307801]

Supplier: R & D Systems
MAB2119
220019-606EA 647.88 USD
220019-606
Anti-GPC3 Mouse Monoclonal Antibody [clone: 307801]
Antibodies
Type: Primary
Antigen Symbol: GPC3
Antigen Name: glypican 3
Antigen Synonyms: DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1
Clonality: Monoclonal
Clone: 307801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR