Anti-EFNA1 Rat Monoclonal Antibody [clone: 93036]

Supplier: R & D Systems
MAB2116
220019-605EA 763.57 USD
220019-605
Anti-EFNA1 Rat Monoclonal Antibody [clone: 93036]
Antibodies
Type: Primary
Antigen: EFNA1
Clonality: Monoclonal
Clone: 93036
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR