Anti-LTA Mouse Monoclonal Antibody [clone: 5802]

Supplier: R & D Systems
MAB211
220019-603EA 779.48 USD
220019-603
Anti-LTA Mouse Monoclonal Antibody [clone: 5802]
Antibodies
Type: Primary
Antigen: LTA
Clonality: Monoclonal
Clone: 5802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR