Anti-IL27RA Rat Monoclonal Antibody [clone: 263503]

Supplier: R & D Systems
MAB21091
220019-602EA 594.37 USD
220019-602
Anti-IL27RA Rat Monoclonal Antibody [clone: 263503]
Antibodies
Type: Primary
Antigen: IL27RA
Clonality: Monoclonal
Clone: 263503
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR