Anti-IL27RA Rat Monoclonal Antibody [clone: 263517]

Supplier: R & D Systems
MAB2109
220019-601EA 577.02 USD
220019-601
Anti-IL27RA Rat Monoclonal Antibody [clone: 263517]
Antibodies
Type: Primary
Antigen: IL27RA
Clonality: Monoclonal
Clone: 263517
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR