Anti-SEMA6C Rat Monoclonal Antibody [clone: 256631]

Supplier: R & D Systems
MAB2108
220019-600EA 763.57 USD
220019-600
Anti-SEMA6C Rat Monoclonal Antibody [clone: 256631]
Antibodies
Type: Primary
Antigen: SEMA6C
Clonality: Monoclonal
Clone: 256631
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR