Anti-VIM Rat Monoclonal Antibody [clone: 280622]

Supplier: R & D Systems
MAB21051
220019-599EA 577.02 USD
220019-599
Anti-VIM Rat Monoclonal Antibody [clone: 280622]
Antibodies
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 280622
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR