Anti-VIM Rat Monoclonal Antibody [clone: 280618]

Supplier: R & D Systems
MAB2105
220019-598EA 647.88 USD
220019-598
Anti-VIM Rat Monoclonal Antibody [clone: 280618]
Antibodies
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 280618
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR