Anti-PROK1 Mouse Monoclonal Antibody [clone: 239727]

Supplier: R & D Systems
MAB2100
220019-595EA 763.57 USD
220019-595
Anti-PROK1 Mouse Monoclonal Antibody [clone: 239727]
Antibodies
Type: Primary
Antigen: PROK1
Clonality: Monoclonal
Clone: 239727
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR