Anti-REG2 Rat Monoclonal Antibody [clone: 239237]

Supplier: R & D Systems
MAB2098
220019-593EA 772.25 USD
220019-593
Anti-REG2 Rat Monoclonal Antibody [clone: 239237]
Antibodies
Type: Primary
Antigen: REG2
Clonality: Monoclonal
Clone: 239237
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR