Anti-SMAD4 Mouse Monoclonal Antibody [clone: 253327]

Supplier: R & D Systems
MAB20971
220019-592EA 577.02 USD
220019-592
Anti-SMAD4 Mouse Monoclonal Antibody [clone: 253327]
Antibodies
Type: Primary
Antigen: SMAD4
Clonality: Monoclonal
Clone: 253327
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR