Anti-SMAD4 Mouse Monoclonal Antibody [clone: 253343]

Supplier: R & D Systems
MAB2097
220019-591EA 594.37 USD
220019-591
Anti-SMAD4 Mouse Monoclonal Antibody [clone: 253343]
Antibodies
Type: Primary
Antigen: SMAD4
Clonality: Monoclonal
Clone: 253343
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR