Anti-SEMA6D Mouse Monoclonal Antibody [clone: 257510]

Supplier: R & D Systems
MAB2095
220019-589EA 594.37 USD
220019-589
Anti-SEMA6D Mouse Monoclonal Antibody [clone: 257510]
Antibodies
Type: Primary
Antigen: SEMA6D
Clonality: Monoclonal
Clone: 257510
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR