Anti-SEMA6B Mouse Monoclonal Antibody [clone: 254402]

Supplier: R & D Systems
MAB2094
220019-588EA 772.25 USD
220019-588
Anti-SEMA6B Mouse Monoclonal Antibody [clone: 254402]
Antibodies
Type: Primary
Antigen: SEMA6B
Clonality: Monoclonal
Clone: 254402
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR