Anti-IL9 Rabbit Monoclonal Antibody [clone: 1043G]

Supplier: R & D Systems
MAB2091
102885-428EA 594.37 USD
102885-428
Anti-IL9 Rabbit Monoclonal Antibody [clone: 1043G]
Antibodies
Type: Primary
Antigen: IL9
Clonality: Monoclonal
Clone: 1043G
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR