Anti-REG1B Mouse Monoclonal Antibody [clone: 293242]

Supplier: R & D Systems
MAB2090
220019-587EA 566.89 USD
220019-587
Anti-REG1B Mouse Monoclonal Antibody [clone: 293242]
Antibodies
Type: Primary
Antigen: REG1B
Clonality: Monoclonal
Clone: 293242
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR