Anti-LYVE1 Mouse Monoclonal Antibody [clone: 537022]

Supplier: R & D Systems
MAB20893
220019-586EA 763.57 USD
220019-586
Anti-LYVE1 Mouse Monoclonal Antibody [clone: 537022]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: LYVE1
Clonality: Monoclonal
Clone: 537022
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR