Anti-LYVE1 Mouse Monoclonal Antibody [clone: 264725]

Supplier: R & D Systems
MAB20891
220019-584EA 779.48 USD
220019-584
Anti-LYVE1 Mouse Monoclonal Antibody [clone: 264725]
Antibodies
Type: Primary
Antigen: LYVE1
Clonality: Monoclonal
Clone: 264725
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR